Imprimer

Indo-1 AM 96 HPLC_ASSAY_METHOD fluorescent indicator, Calcium determination

Marques : Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): 1-[2-Amino-5-(6-carboxyindol-2-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethylester, Indo-1-(acetoxymethyl) ester, 4-(6-Carboxy-2-indolyl)-4′-methyl-2,2′-(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, Indo-1 AM Ester

50044 50043 50043-1
50044.EA 293 EUR
50044. 50043. 50043-1
Indo-1 AM 96 HPLC_ASSAY_METHOD fluorescent indicator, Calcium determination
Indo-1 AM

Indo-1, AM ester is membrane-permeant and thus can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate cell loading. Indo-1-AM Ester itself does not bind Ca2+ , but it is readily hydrolyzed to indo-1 by endogenous esterases once it enters cells.


  • Absorption/Emission 356 nm/478 nm (no Ca2+); 330 nm/400 nm (high Ca2+)
  • Extinction Coefficient 33,000 M -1 cm -1 (no Ca2+); 33,000 M -1 cm -1 (high Ca2+)
  • Cell permeability: Membrane permeant
  • Light yellow solid soluble in DMSO
  • Store desiccated at -20°C and protect from light, especially in solution

Formule: C₄₇H₅₁N₃O₂₂
Poids moléculaire: 1009.91 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00036955
Numéro CAS: 112926-02-0

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR