Imprimer
VWR®, Automated Cell Counters
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
49862-2000EA 3850 EUR
49862-2000 49893-2000 49893-4950 49893-4951 49893-4952
VWR®, Automated Cell Counters
Compteurs de cellules et cytomètres de flux Compteurs de cellules automatisés
Save time and reduce variability with VWR® automated cell counter and VWR® automated cell counter Fluo. Intelligent image recognition and advanced optical imaging technology to achieve accurate and automatic cell analysis, cell concentration and cell size determination.
  • Wide application: cell counting, cell viability, AO/PI staining, cell transfection efficiency, cell expression
  • All-in-one machine with a compact design
  • Dual channel option in VWR® automated cell counter Fluo
  • 3 interchangeable fluorescent cubes (green, blue, red)
  • Cell counting slide
En savoir plus