Imprimer
Bacs de stérilisation, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
6900-0010 6900-0020
489-2120EA 57.2 EUR
489-2120 489-2121
Bacs de stérilisation, Nalgene®
Plateaux/Récipients/Pots Bacs de stérilisation
PP
  • Rolled edges for extra stiffness
  • Handles for easy carrying
For drying and sterilising instruments and labware, collecting and washing soiled lab equipment.