Imprimer
HPLC gradient system diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Marques : Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
48271
48271.EA 101 EUR
48271.
HPLC gradient system diagnostics mix, in methanol: water (3:2) (varied concentration), certified reference material, Supelco®
Étalons Étalons de chromatographie gazeuse
This formulation is designed for evaluating how reliably a chromatographic system is providing such fundamentally important parameters as flow rate, proportioning, and mixing.
Analytes:
Butyl 4-hydroxybenzoate; Ethyl 4-hydroxybenzoate; Heptyl paraben; Methyl 4-hydroxybenzoate; Phenol; Propyl 4-hydroxybenzoate; Uracil
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.