Imprimer
All-day cooling and freezing workstations, Corning® CoolBox™ XT
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
432021 432022 432023 432024 432025 432026 432027 432028
479-1863EA 696 EUR
479-1863 479-1864 479-1865 479-1866 479-1921 479-1922 479-1923 479-1924
All-day cooling and freezing workstations, Corning® CoolBox™ XT
Refroidisseurs Refroidisseurs de paillasses
Ces boîtes sont dédiées au transfert des échantillons entre les laboratoires ou sur le terrain.
  • Over 16 hours of 0,5 to 4 °C cooling with lid closed and over 10 hours with the lid open
  • Reduces contamination by eliminating ice
  • Easy to clean with lab detergents or bleach
  • Patent-pending dual phase conductive XT Cooling core ensures stable and consistent sample cooling
  • Portable XT CoolBox systems can be used when a brief transfer of temperature sensitive samples to a robotic device is needed
  • May be used with dry ice in place of the Cooling Core to create a
En savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable