Imprimer
Desiccators
Fournisseur: SMITH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
262D/250 262D/K/200
SMIT262D/250EA 1050 EUR
SMIT262D/250 467-0031
Desiccators
Dessiccateurs Dessiccateurs en verre
Simax borosilicate glass. Novus pattern, vacuum or knob lid type with tubular in centre of lid. Cover ground to fit.
  • Spare lids are available for these desiccators
Informations de commande: Supplied with lid, removable stopcock (vacuum type only) and perforated ceramic disc.