Imprimer

Copper 65, 65 Cu, plasma standard solution, 65 Cu 10 µg/ml, Specpure®

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

45276.ADEA 185 EUR
45276.AD
Copper 65, 65 Cu, plasma standard solution, 65 Cu 10 µg/ml, Specpure®
Étalons Étalons cationiques Standards de cuivre

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00144006

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR