Imprimer

Buffer HF-Improved

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

44627.AK 44627.K2
44627.AKEA 45.3 EUR
44627.AK 44627.K2
Buffer HF-Improved
Tampons Tampons pH

Buffer used for pretreatment of planar silicon devices when plating with nickel plating solution products 44069 and 44070.


Density: 1.12
Appearance: Colorless
Odor: Acrid
Boiling Point: 103…105°
UN No.: 2817

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR