Imprimer
Magnetic stirring bar arrester, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
F377860000
442-0315EA 80.4 EUR
442-0315
Magnetic stirring bar arrester, SP Bel-Art
Agitateurs Outils d'agitation
Polyethylene (9,5 mm) with radial magnet (Ø 51 mm). Position the arrester against the vessel bottom.
  • Prevents the stirring bar from being poured out with the liquid from a beaker or flask