Imprimer

n-Hexane, anhydre

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Hexyl hydride, Hexanen, Hexane

43263.K2 43263.AK
43263.K2EA 193 EUR
43263.K2 43263.AK
n-Hexane, anhydre
n-Hexane

CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520


Solvent in Sol-Gel precursor applications

Formule: H₃C(CH₂)₄CH₃
Poids moléculaire: 86.18 g/mol
Point d'ébullition: 69 °C
Point de fusion: –95 °C
Densité: 0.659
Point éclair: –23 °C (–9 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00009520
Numéro CAS: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Indice Merck: 14,4694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR