Imprimer

Lithium standard dissolved in oil, 5,000 µg/g Li in mineral oil Specpure® for wear metal analysis

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

42442.1Y 42442.18
42442.1YEA 105 EUR
42442.1Y 42442.18
Lithium standard dissolved in oil, 5,000 µg/g Li in mineral oil Specpure® for wear metal analysis
Étalons Étalons cationiques Standards de lithium

Physical form: liquid
MDL No.: MFCD00134051

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR