Imprimer

Potassium fluorure 99,9 Metals basis

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Fluorure de potassium

42217.14
42217.14EA 343 EUR
42217.14
Potassium fluorure 99,9 Metals basis
Potassium fluorure

Purity: 99.9% CAS No.: 7789-23-3 Molecular Formula: FK Formula Weight: 58.10 Physical Form: Crystalline Density: 2.48 Refractive Index: 1.363 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 859° Boiling Point: 1505° Beilstein Reference: 3902818 Merck Reference: 14,7632 UN No.: 1812 MDL No.: MFCD00011398

Formule: KF
Poids moléculaire: 58.1 g/mol
Point d'ébullition: 1505 °C
Point de fusion: 859 °C
Densité: 2.48
Température de stockage: Température ambiante
Numéro MDL: MFCD00011398
Numéro CAS: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1 III
Indice Merck: 14,7632

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR