Imprimer

Antimony standard solution, 1,000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (de Sb) Specpure® étalon mono-élément pour ICP

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

41682.AD 41682.AE 41682.AP
41682.ADEA 70 EUR
41682.AD 41682.AE 41682.AP
Antimony standard solution, 1,000 mg/L Sb in water with nitric acid and tartaric acid (each max. 1%) (de Sb) Specpure® étalon mono-élément pour ICP
Étalons Étalons cationiques Standards d'antimoine

Molecular Formula: Sb in H₂O/tartaric acid/tr. HNO₃
Physical Form: Liquid
MDL No.: MFCD00134030


Certifications: Traçabilité NIST

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR