Imprimer

Acide sulfurique 2.5 mol/l (5 N) aqueous solution, Titripur® Volumetric, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger


MQ100

4.80364.1000 4.80364.9025
4.80364.1000EA 37 EUR
4.80364.1000 4.80364.9025
Acide sulfurique 2.5 mol/l (5 N) aqueous solution, Titripur® Volumetric, Supelco®
Acide sulfurique

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.

Formule: H₂SO₄
Poids moléculaire: 98.08 g/mol
Point d'ébullition: 330
Point de fusion: 10.38
Densité: 1.146 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064589
Numéro CAS: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR