Imprimer
pH tester, Checker
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
39771.KT
39771.KTEA 128 EUR
39771.KT
pH tester, Checker
Compteurs électrochimiques pH-mètres et multi-paramètres
This pocket size pH meter is supplied with a rugged, epoxy-body pH electrode.
  • Large and easy to read LCD
  • High accuracy with 0,01 pH resolution
  • Two point fast and accurate calibration
  • Secure single action on/off switch
  • Usable with virtually any pH electrode
  • Battery life: 3000 hours
Informations de livraison: Suplied with screw-type, epoxy-body pH electrode.