Imprimer
VWR®, Cell culture salt solutions, Dulbecco's Phosphate Buffered Saline (DPBS)
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
392-0433EA 14.1 EUR
392-0433 392-0434 392-0440
VWR®, Cell culture salt solutions, Dulbecco's Phosphate Buffered Saline (DPBS)
Tampons Tampons de culture de tissus
DPBS is commonly used salt solution in cell enumeration as a diluent, for rinsing cells and as a buffer in many chromatographic procedures. DPBS is also used in FACS (Fluorescence-Activated Cell Sorting) process to wash and re-suspend cells during the dissociation process, as well as for many analytical methods or biochemical treatments.
For applications where Ca²⁺ and Mg²⁺ ions interfere with enzyme activity (e.g, Trypsin), DPBS is:
- Without calcium
- Without magnesium