Imprimer
Temperature indicating tubes
Fournisseur: KOEHLER TECHNISCHE PRODUKTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
K-1253 K-1252 K-1250
391-0084EA 232 EUR
391-0084 391-0085 KOEHK-1250
Temperature indicating tubes
Bandelettes indicatrices de température
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.