Imprimer
Peroxidase Indicator Reagent
Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3901-10VL
3901-10VLEA 382 EUR
3901-10VL
Peroxidase Indicator Reagent
Réactifs d'électrophorèse Western Blotting Reagents
It contains p-Phenylenediamine diHCL (1 part) and catechol (2 parts). Commonly referred to as Hanker Yates reagent. Kit component in the Sigma Leukocyte peroxidase (myeloperoxidase) Procedure, 390A.