Imprimer
Constant Climate Chambers, KBF Series
Fournisseur: Binder
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
9020-0478 9020-0479 9020-0480 9020-0481 9020-0482 9020-0483
390-1753EA 17440 EUR
390-1753 390-1754 390-1755 390-1756 390-1757 390-1758
Constant Climate Chambers, KBF Series
Chambres d'essai Chambres climatiques
The KBF series is a proven specialist for reliable ICH stability tests in the pharmaceutical industry: the KBF masters all long-term tests, accelerated tests and stress tests. It fulfills all important ICH requirements, from precision to programming and documentation. With flexible water supply options for all installation locations. The units have a stainless-steel inner chamber and proven APT.line™ preheating chamber technology, feature a temperature range of 0 to +70 °C, and humidity range of 10 to 80% RH, the KBF consumes up to 40 % less energy than its predecessor. With optionalEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Modèle KBF 130 KBF 260 KBF 470 KBF 720 KBF 1060 KBF 1600
Gamme de température 0...70 °C (without humidity),
10...70 °C (with humidity)
0...70 °C (without humidity),
10...70 °C (with humidity)
0...70 °C (without humidity),
10...70 °C (with humidity)
0...70 °C (without humidity),
10...70 °C (with humidity)
0...70 °C (without humidity),
10...70 °C (with humidity)
0...70 °C (without humidity),
10...70 °C (with humidity)
Environmentally Preferable
Stabilité de température (temps) ±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
±0.1 K (at 25 °C and 60% RH)
±0.1 K (at 40 °C and 75% RH)
Homogénéité de température ±0.2 K (at 25 °C and 60% RH)
±0.2 K (at 40 °C and 75% RH)
±0.3 K (at 25 °C and 60% RH)
±0.2 K (at 40 °C and 75% RH)
±0.2 K (at 25 °C and 60% RH)
±0.2 K (at 40 °C and 75% RH)
±0.3 K (at 25 °C and 60% RH)
±0.2 K (at 40 °C and 75% RH)
±0.3 K (at 25 °C and 60% RH)
±0.2 K (at 40 °C and 75% RH)
±0.4 K (at 25 °C and 60% RH)
±0.3 K (at 40 °C and 75% RH)
Gamme d'humidité (HR) 10...80 % 10...80 % 10...80 % 10...80 % 10...80 % 10...80 %
Capacité 127 L 262 L 472 L 727 L 1070 L 1610 L
Type de convection Convection forcée Convection forcée Convection forcée Convection forcée Convection forcée Convection forcée
Plateaux livrés (max.) 2 (6) 2 (9) 2 (16) 2 (16) 2 (16) 2 (16)
Charge maximale par clayette 25 kg 30 kg 40 kg 40 kg 40 kg 40 kg
Puissance nominale 1.3 kW 1.4 kW 1.4 kW 1.6 kW 1.6 kW 1.7 kW
Poids 126 kg 188 kg 225 kg 298 kg 340 kg 438 kg