Imprimer
Heracell Vios CR CO₂ incubators, CTS series, Thermo Scientific™
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
51033772 51033775 51033784 51033787
390-1576EA 20610 EUR
390-1576 390-1577 390-1578 390-1579
Heracell Vios CR CO₂ incubators, CTS series, Thermo Scientific™
Incubateurs Incubateurs CO₂
Heracell Vios CR cleanroom compatible CO₂ incubators are third party-certified, suitable for use in ISO Class 5 and GMP Grade A/B environments.
  • Next level particulate and contamination control
  • Active particle control system captures particulates to help assure continued cleanroom compatibility
  • Sealed exterior casing with stainless steel interior and exterior
  • IP 54 rated electronics, protected from dust and spray in any direction
  • Thermo Scientific™ THRIVE™ active airflow technology delivers homogeneous growth conditions quickly
  • IR sensor and proven 180 °C
En savoir plus