Imprimer
Cooled Incubators with Advanced Peltier Technology, IPP and IPPeco series
Fournisseur: MEMMERT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
IPP30 IPP30PLUS IPP55 IPP55PLUS IPP1060ECO IPP1060ECOPLUS IPP110ECO IPP110ECOPLUS IPP1400ECO IPP1400ECOPLUS IPP2200ECO IPP2200ECOPLUS IPP260ECO IPP260ECOPLUS IPP410ECO IPP410ECOPLUS IPP750ECO IPP750ECOPLUS
390-0622EA 3530 EUR
390-0622 390-0623 390-0624 390-0625 390-1416 390-1459 390-1470 390-1493 390-1504 390-1505 390-1506 390-1507 390-1508 390-1509 390-1512 390-1513 390-1514 390-1515
Cooled Incubators with Advanced Peltier Technology, IPP and IPPeco series
Incubateurs Incubateurs réfrigérés
Sets eco standards for cultivation below room temperature: Unmatched energy efficiency, excellent heat-up, cool-down and recovery times.
  • Maximum energy efficiency compared to cooled incubators with compressor
  • Completely homogenous distribution of temperature
  • Wider range of applications thanks to cooling down to the freezpoint
  • Refrigerant-free and low-maintenance
These reliable Peltier-cooled incubators are available in nine different sizes (30, 55, 110, 260, 410, 750, 1060, 1400 and 2200 L). Perfectly coordinated systems and excellent efficiency makeEn savoir plus