Imprimer

Ammonium sulfate 99.0-100.5%, granuleux FCC, purifié, Macron Fine Chemicals™

Marques : Avantor Performance Materials
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

3496-28EA 0 EUR
3496-28
Ammonium sulfate 99.0-100.5%, granuleux FCC, purifié, Macron Fine Chemicals™
Ammonium sulfate
Formule: (NH₄)₂SO₄
Poids moléculaire: 132.14 g/mol
Densité: 1.77 g/cm³
Numéro MDL: MFCD00003391
Numéro CAS: 7783-20-2
Indice Merck: 13,00559

Commander maintenant

Résultats des tests de spécification

Meets F.C.C. Requirements
Food GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical 99.0 - 100.5 %
FCC - Assay ((NH₄)₂SO₄) 99.0 - 100.5 %
FCC - Arsenic (As) ≤3 ppm
FCC - Identification A Passes Test
FCC - Identification B Passes Test
FCC - Iron (Fe) ≤0.0010 %
FCC - Lead (Pb) ≤3 mg/kg
FCC - pH of Aqueous Solution 4.5 - 6.0
FCC - Residue after Ignition ≤0.25 %
FCC - Selenium (Se) ≤0.0030 %
Solubility Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR