Imprimer
Flacons à col large, PEHD, économiques, avec bouchon à vis, Nalgene®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
DS2185-0004 DS2185-0016 DS2185-0032 DS2185-0008
331-4213EA 137 EUR
331-4213 331-4215 NALGDS2185-0032 331-0300
Flacons à col large, PEHD, économiques, avec bouchon à vis, Nalgene®
Flacons Flacons en plastique
Wide mouth amber bottles made up of HDPE designed to protect the light sensitive liquids. It is used for storage, shipping and packaging.
  • Economical and recyclable
  • Designed for single use
  • Chemical resistance to most acids, bases and alcohols
  • Can be used in freezer to −100 °C for long term storage
  • Linerless caps ensure leakproof performance
These bottles are for single use and non autoclavable.
Certifications: Compliance with U.S. Pharmacopoeia 23 requirements for maximum light transmission.
Informations de livraison: Bottles are supplied withEn savoir plus