Imprimer

Diphényl-1,5-carbazide ACS

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): 1,5-Diphénylcarbonohydrazide

31390.06 31390.14 31390.22
31390.06EA 37.8 EUR
31390.06 31390.14 31390.22
Diphényl-1,5-carbazide ACS
Diphényl-1,5-carbazide

CAS No.: 140-22-7 Molecular Formula: C13H14N4O Formula Weight: 242.28 Physical Form: Crystalline Sensitivity: Air and Light Sensitive Appearance: White to cream Melting Point: 170-176° Solubility: Soluble in hot alcohol, acetone, glacial acetic acid Beilstein Reference: 752039 Merck Reference: 14,3321 MDL No.: MFCD00003013

Formule: C₁₃H₁₄N₄O
Poids moléculaire: 242.28 g/mol
Point d'ébullition: 454
Point de fusion: 170…176 °C
Densité: 1.31 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00003013
Numéro CAS: 140-22-7
EINECS: 205-403-7
Indice Merck: 14,3321
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR