Imprimer
Spray réfrigérant
Fournisseur: HM INDUSTRIESERVICE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
100089
291-1203EA 51.9 EUR
291-1203
Spray réfrigérant
Bombes réfrigérantes
For applications in the vicinity of ignition sources or in potentially explosive areas. Used to detect thermal faults in electronics, to join small parts by means of cold shrinkage, to remove sticky dirt from textiles, to quick-freeze rubber and plastic and to test the function of thermostats and temperature probes.
  • Cools down to −55 °C
  • No risk of ignition or explosion, even in the presence of ignition sources
  • Does not contain any CFCs