Imprimer
Affinity chromatography columns, antibody package, protein A
Fournisseur: Cytiva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
29-0588-05
29-0588-05EA 191 EUR
29-0588-05
Affinity chromatography columns, antibody package, protein A
Colonnes pour chromatographie
This Protein A starter pack includes a prepacked protein A affinity column and a prepacked desalting column for routine purification of antibodies such as human IgG.
  • HiTrap™ Desalting column is included for quick pH neutralisation of eluted antibodies
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as ÄKTA system
HiTrap™ Protein A HP is prepacked with Protein A Sepharose™ High Performance. This resin is used for routine immunoaffinity purifications of most IgG from ascites, serum, and cellEn savoir plus