Imprimer

HiScreen Phenyl FF (High Sub) columns

Fournisseur: Cytiva
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

28-9269-88
28-9269-88EA 166 EUR
28-9269-88
HiScreen Phenyl FF (High Sub) columns
Colonnes pour chromatographie
Phenyl Sepharose 6 Fast Flow is a well established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.Two levels of substitution help find optimal selectivity and binding capacity for a given application.

  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Prepacked, ready-to-use columns for convenient process development optimization
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and
En savoir plus
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR