Imprimer

Thrombin

Marques : Cytiva
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

27-0846-01
27-0846-01EA 177 EUR
27-0846-01
Thrombin
Enzymes

Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors containing the recognition sequence for thrombin (pGEX-1lT, pGEX-2T, pGEX-2TK, pGEX-4T-1, pGEX-4T-2, and pGEX-4T-3).


  • Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
  • Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose™ High Performance, or Glutathione Sepharose™ 4 Fast Flow


Application: Used for site-specific separation of the GST tag from proteins expressed using pGEX-T vectors.


Cleavage Buffer: PBS 140 mM NaCl, 2,7 mM KCl, 10 mM Na₂HPO₄, 1,8 mM KH₂PO₄, pH 7,3
Elution buffer: 50 mM Tris-HCl, 10 mM reduced glutathione, pH 8,0.


Inhibitors: Pefabloc TH benzamidine, AEBSF, APMSF, antithrombin III, Antipain, α1-antitrypsin, aprotinin, chymostatin, hirudin, leupeptin, PMSF.


Molecular weight: 37000.


Unit definition: One unit of enzyme cleaves 90% of 100 μg of a test GST fusion protein when incubated in 1X PBS at 22 °C for 16 hours.


Informations de commande: Supplied as lyophilised powder.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR