Imprimer

Peqlab peqGOLD, DNA Ladder, 100 bp Plus

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

25-2020EA 76.2 EUR
25-2020
Peqlab peqGOLD, DNA Ladder, 100 bp Plus
Réactifs d'électrophorèse Marqueurs de poids moléculaires

peqGOLD DNA ladder 100 bp plus is ideal for size determination in the range of 100 to 3000 base pairs. There are no unspecific bands besides the fragments described. 1000 and 500 bp bands have a higher DNA content and serve as a size reference.


  • Composed of 14 chromatography-purified individual DNA fragments: 3000, 2000, 1500, 1200, 1000, 900, 800, 700, 600, 500, 400, 300, 200 and 100 base pairs


Informations de livraison: Product comprises of 100 µl (50 µg) DNA ladder and 1 ml 6X loading buffer (10 mM Tris-HCl, pH 7,6; 0,03% bromophenol blue; 0,03% xylene cyanol; 60% glycerol and 60 mM EDTA)


Attention: The marker has to be dephosphorylated before being used in radioactive assays.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR