Imprimer
Élévateurs de laboratoire, Swiss Boy
Fournisseur: GRAUER RUDOLF
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
100 105 110 115 116 116MS 118 122 122R 122MR
245-1110EA 270 EUR
245-1110 245-1120 245-1130 245-1140 245-1150 245-1152 245-1160 245-1170 245-1171 245-1173
Élévateurs de laboratoire, Swiss Boy
Crics de laboratoire
These laboratory jacks are made of anodised aluminium for corrosion resistance and resistance to many chemicals.
  • Easy adjustment with resilience
  • High stability and high load carrying capacity
The motorised models type 116 and 122 feature a variable adjustment of the lower and upper end positions. When the set position has been reached the lab jack stops automatically. These models are also suitable for repetitive operations.