Imprimer
Separating funnel holder
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
140300
241-0345EA 12.4 EUR
241-0345
Separating funnel holder
Supports de statif
PP, white.
For conical separating funnels up to 1000 ml. Easely mounted on support rods with an outer Ø 8 to 14 mm with a locking nut.