Imprimer
Capillary columns, special purpose, FAMES, SP-2560, Supelco®
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
24056
24056.EA 2550 EUR
24056.
Capillary columns, special purpose, FAMES, SP-2560, Supelco®
Colonnes pour chromatographie
Specially prepared and tested columns to seperate geometrical position (cis/trans) isomers of fatty acid methyl esters. Seperation of FAMEs should be take place in hydrogenated vegetable oil samples.
Temperature limits: Subambient to 250 °C
Phase: Nonbonded and biscyanopropyl polysiloxane