Imprimer

Ethyle acétate 99,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Acétate d'éthyle

23882.296EA 80.5 EUR
23882.296 23882.321 23882.467 VWRC23882.550 23882.330 23882.310 23882.360 23882.950
Ethyle acétate 99,5, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Ethyle acétate
Formule: CH₃COOC₂H₅
Poids moléculaire: 88.11 g/mol
Point d'ébullition: 77.1 °C (1013 hPa)
Point de fusion: –83 °C
Densité: 0.902 g/cm³ (20 °C)
Point éclair: –4 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00009171
Numéro CAS: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Indice Merck: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (on anhydrous substance) Min. 99.5 %
IR Spectrum Passes test
Organic impurities Passes test
Acidity Max. 0.0006 meq/g
Boiling point 76.0 - 77.5 °C
Colouration Max. 10 APHA
Density (20/4) 0.899 - 0.901
Density (20/20) 0.901 - 0.904
Substances coloured by H2SO4 Max. 60 APHA
Ethanol Max. 0.1 %
Evaporation residue Max. 10 ppm
Methanol Max. 0.1 %
Methyl acetate Max. 0.1 %
iso-Propyl acetate Max. 0.06 %
n-Propyl acetate Max. 0.5 %
Water Max. 0.03 %
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.05 ppm
Co (Cobalt) Max. 0.02 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.1 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPÉCIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR