Imprimer
Sampling spatula, Sterileware®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
H369300000 H369300050
231-0014EA 463 EUR
231-0014 BELAH369300050
Sampling spatula, Sterileware®, SP Bel-Art
Spatules
One-piece spatulas, moulded from FDA grade, high impact PS.
  • Sterilised by gamma irradiation, lot stamped for accurate lot tracing, shipped with a Certificate of Processing for Sterilisation
V-shaped spatula for sampling dislodge caked or crystallised reagents from deep within tall bottles or vessels having a neck diameter as small as 18 mm (³/₄"). The sturdy point is perfect for taking sterile samples during process quality inspections, and for piercing plastic-lined paper bags or similar packages.