Imprimer

n-Pentane 99 HPLC

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Pentane

Filter 0,2 um

22907.K2 22907.M1
22907.K2EA 96.1 EUR
22907.K2 22907.M1
n-Pentane 99 HPLC
n-Pentane

Purity: 99% min CAS No.: 109-66-0 Molecular Formula: CH3(CH2)3CH3 Formula Weight: 72.15 Flash Point: -49°(-57°F) Physical Form: Liquid Density: 0.626 Refractive Index: 1.3580 Appearance: Colourless Melting Point: -130° Boiling Point: 36° Merck Reference: 14,7116 UN No.: 1265 MDL No.: MFCD00009498

Formule: H₃C(CH₂)₃CH₃
Poids moléculaire: 72.15 g/mol
Point d'ébullition: 36 °C
Point de fusion: –130 °C
Densité: 0.626
Point éclair: –49 °C (–57 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00009498
Numéro CAS: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3 II
REACH: 01-2119459286-30
Indice Merck: 14,7116
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance (Color) Colorless
Form Liquid
Color (APHA) ≤10
Assay (GC) ≥99.9% ( As C5 Hydrocarbons)
Comment Sub micron filtered
Water Content (Karl Fischer Titration) ≤0.02%
Optical Absorbance at 200 nm ≤1.0 AU
Optical Absorbance at 210 nm ≤0.5 AU
Optical Absorbance at 220 nm ≤0.15 AU
Optical Absorbance at 245 nm ≤0.01 AU
Residue on Evaporation ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR