Imprimer

Chloroforme 99.0-99.4% stabilisé, AnalaR NORMAPUR® Reag. Ph. Eur., ACS pour analyses

Marques : VWR Chemicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): Trichlorométhane

Stabilisation: 0.4-1 % Stabilisé avec ethanol

22711.324EA 209 EUR
22711.324 22711.244 22711.461 22711.290 22711.260 22711.540 22711.420
Chloroforme 99.0-99.4% stabilisé, AnalaR NORMAPUR® Reag. Ph. Eur., ACS pour analyses
Chloroforme
Formule: CHCl₃
Poids moléculaire: 119.38 g/mol
Point d'ébullition: 61.7 °C (1013 hPa)
Point de fusion: –63 °C
Densité: 1.4832 g/cm³ (20 °C)
Température de stockage: Température ambiante
Numéro MDL: MFCD00000826
Numéro CAS: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1 III
REACH: 01-2119486657-20
Indice Merck: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=c7d46a2ee73651eae0f3654c272e820106b2f04f&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Appearance Clear colourless liquid
Assay 99.0 - 99.4 %
Assay (+ stabilizing agent) Min. 99.8 %
Insolubility in alcohol Passes test
IR Spectrum Passes test
Suited for determination with dithizone Passes test
Acidity Max. 0.00014 meq/g
Boiling point 59.5 - 60.5 °C
Colouration Max. 10 APHA
Density (20/4) 1.476 - 1.485
Density (20/20) 1.475-1.481
Substances coloured by H2SO4 Max. 35 APHA
Carbon tetrachloride Max. 100 ppm
Carbonyl compounds (as CO) Max. 50 ppm
Dichloromethane Max. 100 ppm
Ethanol 0.4 - 1.0 %
Evaporation residue Max. 5 ppm
Free chlorine Max. 0.3 ppm
Ionised chlorine Max. 0.5 ppm
Tetrachloroethylene Max. 100 ppm
Trichloroethylene Max. 100 ppm
Water Max. 100 ppm
Cl (Chloride) Max. 0.2 ppm
Al (Aluminium) Max. 0.05 ppm
B (Boron) Max. 0.01 ppm
Ba (Barium) Max. 0.01 ppm
Ca (Calcium) Max. 0.1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.2 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.05 ppm
Conforms to Reag. Ph.Eur. Passes test
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR