Imprimer
Specimen jars with knobbed drop-on cover
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
632416130808 632416131212 632416131515 632416132020
216-3351EA 76.5 EUR
216-3351 216-3353 216-3354 216-3355
Specimen jars with knobbed drop-on cover
Pots Pots de spécimens
Made of glass with optical purity.
  • Sturdy
  • Excellent chemical resistance
  • Does not disfigure the view of content
Specimen jars with base and drop-on cover are suitable for use in biology and medicine for specimen storage.