Imprimer
Ointment jars, with thread
Fournisseur: RIXIUS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1-0900-005051I 1-0900-0030-46 1-0900-0100-62 1-0901-0100-52
216-2983EA 90.5 EUR
216-2983 216-2972 216-2981 216-2979
Ointment jars, with thread
Pots Pots à usage général
Made from clear or amber soda-lime glass.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • With round thread
Ideal for sampling, storage and transportation of powders and pastes. Amber jars to protect light-sensitive materials.
Informations de commande: Screw caps have to be ordered separately.