Imprimer
Dishes, flat bottom, with spout
Fournisseur: Agencinox
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
312A 316A 320A 325A 306 308 310 312 304
216-2715EA 56.4 EUR
216-2715 216-2716 216-2717 216-2718 216-2711 216-2712 216-2713 216-2714 AGEN304
Dishes, flat bottom, with spout
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Stainless steel 18/10.
  • Without handles
  • Good chemical resistance
  • Excellent thermal resistance
Round multipurpose dishes for a wide range of uses.