Imprimer
Vydac 214TP C4, HPLC columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
554-7580EA 1220 EUR
554-7580 214FSK510 214GCC510 214TP5205 214TP5405 214TP5210 214TP5410 214TP5115 214TP5215 214TP5315 214TP5415 214TP51 214TP52 214TP53 214TP510 214TP10415 214TP101015 214TP104 214TP1010 214TP1022 214GCC1210 214FSK1210 214FSK1510 214TP10152215 214TP101510 214TP101522 214GCC152010 214TP152022 214GK104/N 214GK52/N 214GK54/N 214GD104/N 214GD34/N 214GD52/N 214GD54/N 214GCC1520/N 214GCC1010 HICH5138110 214GD51/N 214GK51/N
Vydac 214TP C4, HPLC columns
Colonnes pour chromatographie
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins,En savoir plus