Imprimer
VWR®, Beakers, Low Form, Heavy-Duty
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
213-0462EA 59.8 EUR
213-0462 213-0476 213-0477 213-0478 213-0479 213-0480 213-0481
VWR®, Beakers, Low Form, Heavy-Duty
Béchers
Borosilicate 3.3 glass, with spout.
  • Good chemical and thermal shock resistance
  • Durable labelling area with matt finish and white graduated scale
  • Spout designed for optimum performance
Heavy-duty beakers have better mechanical stability than the standard products due to thicker walls. The reinforced rims increase shock resistance and reduce the risk of breakage. Due to the increased wall thickness resistance to rapid temperature changes is not as good as that of the standard product, therefore, slow, uniform heating is recommended.