Imprimer
Test tubes, with rim
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
88323 88324 88322 88317 88319 88325
KART88323EA 213 EUR
KART88323 KART88324 KART88322 KART88317 KART88319 212-6894
Test tubes, with rim
Tubes Tubes à essais
PS or PP.
  • Disposable tubes with round or conical bottom
  • With rim
  • For laboratory use
  • Can be used in centrifuges
Certifications: IVD certified.