Imprimer
VWR®, Test Tubes with or without Rim
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
212-0766EA 52.9 EUR
212-0766 212-0767 212-0768 212-0769 212-0770 212-0771 212-0772 212-0773 212-0774 212-0775 212-0776 212-0777 212-0778 212-0779 212-0780 212-0781 212-0782 212-0783 212-0784 212-0785 212-0786 212-0787 212-0788 212-0789
VWR®, Test Tubes with or without Rim
Tubes Tubes à essais
Test tubes made from borosilicate 3.3 glass.
  • Thick walled
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to rapid temperature changes
The economical alternative for a wide range of uses.
Certifications: DIN 12395.