Imprimer

Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

20330
20330.EA 249 EUR
20330.
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Anticorps
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR