Imprimer
Glass stoppers for iodine flasks, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SIB24 SIB29
201-8714EA 102 EUR
201-8714 BIBBSIB29
Glass stoppers for iodine flasks, Quickfit®
Bouchons Bouchons en verre
Quickfit® iodine flask stoppers.
  • Extended handle which projects well above the liquid seal
  • Available with either 24/29 or 29/32 ground cone joints
For use with Quickfit® Iodine flasks.