Imprimer
Air/steam inlet tubes, straight, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MF15/1 MF15/2 MF15/3
201-3056EA 25.2 EUR
201-3056 201-3057 201-3058
Air/steam inlet tubes, straight, Quickfit®
Tubes Tubes d'arrivée de gaz
Pyrex® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Cone with stem and reduced shank
Useful for construction of gas inlet tubes.
Stem length: 150 mm
Three shank lengths: 45/60/70 mm.