Imprimer
Cold trap, Quickfit®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
MF28/52
201-3052EA 134 EUR
201-3052
Cold trap, Quickfit®
Pièges à vide
Designed for the vacuum systems which prevents the vapours from the volatile materials from damaging pumps and easy to use.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable