Imprimer
Distillation apparatus, semi-micro, laboratory set with joints NS14/19
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
11.000
201-0209EA 493 EUR
201-0209
Distillation apparatus, semi-micro, laboratory set with joints NS14/19
Composants de distillation Pièces pour distillation
Box with standard cut-out details for half-scale device assembly.
The kit consists of the following components:
1 teaspoon flask 25 ml NS 19/26
1 teaspoon 50 ml NS 19/26
1 pcs plug NS 14/23
1st Reduction NS 19/26 - NS 14/23
1 st Expansion piece NS 14/23 - NS 19/26
1 st. Dest. NS 19/26 - NS 14/23 - thread GL 18 with screw cap
1 thermometer 0 to 250 °C
1 thermometer 0 to 360 °C
1 piece fastener NS 14/23
1 st cooler 160 mm NS 14/23
1 drip funnel, graduated, 50 ml, NS 14/23
1 pcs grinding clips
This is a basic kit that can be supplemented with single details or complete completion kits. SpecialEn savoir plus