Imprimer

Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

20093 20093-1
20093.EA 359 EUR
20093. 20093-1
Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)
Anticorps
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR