Imprimer
Alkaline cleaning agent, neodisher® LaboClean LA
Fournisseur: Dr. Weigert
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
410681
148-3176EA 365 EUR
148-3176
Alkaline cleaning agent, neodisher® LaboClean LA
Produits nettoyants Détergents et Nettoyants de Surfaces
neodisher® LaboClean LA has an alkaline formulation and it contains high proportions of emulsifiers and dispersing agents. neodisher® LaboClean removes oils, fats, resins, paraffins and organic dyestuffs. At temperature over 30 °C neodisher® LaboClean LA exhibits a defoaming actions.
  • Can be used with any level of water hardness
neodisher® LaboClean LA does not attack stainless steel or precious metals. neodisher® LaboClean is not suitable for aluminium, anodises aluminium and light metal alloys. Borosilicate glass is resistant against neodisher®En savoir plus